4-Chloro-2,5-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16213-25G 16213-5G
76925-682EA 225.98 USD
76925-682 76925-684
4-Chloro-2,5-difluorobenzaldehyde ≥97%
4-Chloro-2,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD07782051
CAS Number: 879093-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR