5-Bromo-2-ethoxynicotinic acid ≥97%

Supplier: AOBChem USA

Warning

16188-1G 16188-25G 16188-5G
76925-670EA 64.72 USD
76925-670 76925-672 76925-674
5-Bromo-2-ethoxynicotinic acid ≥97%
5-Bromo-2-ethoxynicotinic acid
Formula: C₈H₈BrNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09836166
CAS Number: 393184-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR