5-Bromo-3-fluorosalicylaldehyde ≥97%

Supplier: AOBChem USA

Warning

16138-25G 16138-5G
76925-660EA 198.39 USD
76925-660 76925-662
5-Bromo-3-fluorosalicylaldehyde ≥97%
5-Bromo-3-fluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD03788530
CAS Number: 251300-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR