5-Bromo-2-methoxypyridine-3-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-methoxynicotinaldehyde

16109-25G 16109-5G
76925-648EA 453.01 USD
76925-648 76925-650
5-Bromo-2-methoxypyridine-3-carboxaldehyde ≥95%
5-Bromo-2-methoxypyridine-3-carboxaldehyde
Formula: C₇H₆BrNO₂
MW: 216.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663521
CAS Number: 103058-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR