4-Bromo-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15956-1G 15956-25G 15956-5G
76925-600EA 62.6 USD
76925-600 76925-602 76925-604
4-Bromo-2-methoxyphenylboronic acid ≥97%
4-Bromo-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11856025
CAS Number: 889849-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR