5-Bromo-3-chloro-2-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15884-1G 15884-25G 15884-500MG 15884-5G
76925-564EA 224.92 USD
76925-564 76925-566 76925-568 76925-570
5-Bromo-3-chloro-2-hydroxybenzaldehyde ≥97%
5-Bromo-3-chloro-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
MW: 235.462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00807894
CAS Number: 19652-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR