5-Bromo-2-methoxy-4-pyridineboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-2-methoxypyridin-4-yl)boronic acid, 5-Bromo-2-methoxy-4-pyridinylboronic acid, 5-Bromo-2-methoxypyridine-4-boronic acid

15779-1G 15779-25G 15779-5G
76925-528EA 87 USD
76925-528 76925-530 76925-532
5-Bromo-2-methoxy-4-pyridineboronic acid ≥95%
5-Bromo-2-methoxy-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BBrNO₃
Storage Temperature: Ambient
MDL Number: MFCD07781223
CAS Number: 957060-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR