4-Bromo-3-chloro-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

15704-25G 15704-5G
76925-516EA 325.7 USD
76925-516 76925-518
4-Bromo-3-chloro-2-fluoropyridine ≥97%
4-Bromo-3-chloro-2-fluoropyridine
Formula: C₅H₂BrClFN
Storage Temperature: Refrigerator
MDL Number: MFCD11040607
CAS Number: 1017793-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR