5-Bromo-2-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15670-25G 15670-5G
76925-494EA 410.57 USD
76925-494 76925-496
5-Bromo-2-fluoro-3-methylbenzaldehyde ≥97%
5-Bromo-2-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD11110261
CAS Number: 903875-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR