5-Bromo-2-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

15519-25G 15519-5G
76925-460EA 381.93 USD
76925-460 76925-462
5-Bromo-2-(trifluoromethoxy)benzoic acid ≥97%
5-Bromo-2-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₃
MW: 285.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203214
CAS Number: 403646-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR