4-Bromo-3,5-dichlorofluorobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1,3-dichloro-5-fluorobenzene

15205-25G 15205-5G
76925-430EA 325.7 USD
76925-430 76925-432
4-Bromo-3,5-dichlorofluorobenzene ≥97%
4-Bromo-3,5-dichlorofluorobenzene
Formula: C₆H₂BrCl₂F
MW: 243.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845890
CAS Number: 263333-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR