4-Ethoxy-3-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Ethoxy-3-(trifluoromethyl)benzeneboronic acid, 4-Ethoxy-3-trifluoromethylphenylboronic acid

15101-25G 15101-5G
76925-416EA 386.17 USD
76925-416 76925-418
4-Ethoxy-3-(trifluoromethyl)phenylboronic acid ≥97%
4-Ethoxy-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BF₃O₃
MW: 233.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363789
CAS Number: 871329-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR