5-Fluoro-2-(methoxymethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15087-25G 15087-5G
76925-404EA 519.85 USD
76925-404 76925-406
5-Fluoro-2-(methoxymethoxy)benzaldehyde ≥95%
5-Fluoro-2-(methoxymethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD24685864
CAS Number: 603041-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR