5-Carboxy-2-chloro-4-methoxybenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Borono-4-chloro-2-methoxybenzoic acid, 5-Carboxy-2-chloro-4-methoxyphenylboronic acid

15015-1G 15015-25G 15015-500MG 15015-5G
76925-376EA 259.93 USD
76925-376 76925-378 76925-380 76925-382
5-Carboxy-2-chloro-4-methoxybenzeneboronic acid ≥97%
5-Carboxy-2-chloro-4-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BClO₅
Storage Temperature: Ambient
MDL Number: MFCD09878319
CAS Number: 957066-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR