4-Chloro-2-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-formylbenzeneboronic acid

14985-25G 14985-5G
76925-356EA 279.02 USD
76925-356 76925-358
4-Chloro-2-formylphenylboronic acid ≥97%
4-Chloro-2-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₃
MW: 184.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689492
CAS Number: 913835-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR