4-Fluoro-3-iodobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

14910-1G 14910-25G 14910-500MG 14910-5G
76925-340EA 138.98 USD
76925-340 76925-342 76925-344 76925-346
4-Fluoro-3-iodobenzonitrile ≥97%
4-Fluoro-3-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FIN
MW: 247.100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458123
CAS Number: 159719-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR