4-Cyano-2-fluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Cyano-2-fluorophenylboronic Acid, 4-Cyano-2-fluorobenzeneboronic acid

14891-1G 14891-25G 14891-500MG 14891-5G
76925-330EA 171.87 USD
76925-330 76925-332 76925-334 76925-336
4-Cyano-2-fluorophenylboronic acid ≥95%
4-Cyano-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07644472
CAS Number: 1150114-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR