4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Methyl-2-thiopheneboronic acid Pinacol ester, 4-Methylthiophene-2-boronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(4-methylthiophen-2-yl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(4-methyl-2-thienyl)-1,3,2-dioxaborolane

14684-25G 14684-5G
76925-260EA 275.84 USD
76925-260 76925-262
4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97%
4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD11045430
CAS Number: 635305-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR