5-(Methoxycarbonyl)thiophene-2-boronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxylate

14667-1G 14667-25G 14667-500MG 14667-5G
76925-248EA 206.88 USD
76925-248 76925-250 76925-252 76925-254
5-(Methoxycarbonyl)thiophene-2-boronic acid, pinacol ester ≥97%
5-(Methoxycarbonyl)thiophene-2-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₄S
MW: 268.137 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09952057
CAS Number: 916138-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR