4-Ethoxy-2-(trifluoromethyl)phenylboronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(4-Ethoxy-2-(trifluoromethyl)phenyl)boronic acid
14629-25G
14629-5G
76925-238EA
608.96
USD
76925-238
76925-240
4-Ethoxy-2-(trifluoromethyl)phenylboronic acid ≥95%
4-Ethoxy-2-(trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₀BF₃O₃ Storage Temperature: Ambient |
MDL Number:
MFCD08059515 CAS Number: 313545-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...