5-Cyano-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14422-1G 14422-25G 14422-5G
76925-194EA 62.6 USD
76925-194 76925-196 76925-198
5-Cyano-2-methoxyphenylboronic acid ≥97%
5-Cyano-2-methoxyphenylboronic acid
Formula: C₈H₈BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD06801691
CAS Number: 612833-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR