4-Methyl-3-(1-Methylethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

14246-1G 14246-25G 14246-5G
76925-158EA 111.4 USD
76925-158 76925-160 76925-162
4-Methyl-3-(1-Methylethoxy)benzoic acid ≥97%
4-Methyl-3-(1-Methylethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14653014
CAS Number: 1014691-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR