5-Chloro-3-methylpicolinaldehyde ≥95%

Supplier: AOBChem USA

Warning

14198-1G 14198-25G 14198-500MG 14198-5G
76925-142EA 266.29 USD
76925-142 76925-144 76925-146 76925-148
5-Chloro-3-methylpicolinaldehyde ≥95%
5-Chloro-3-methylpicolinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.582 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18258016
CAS Number: 780801-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR