6-Chloro-5-methylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-5-methylpyridine-3-carboxylic acid, 5-Carboxy-2-chloro-3-methylpyridine

14194-1G 14194-25G 14194-500MG 14194-5G
76925-128EA 153.84 USD
76925-128 76925-130 76925-132 76925-134
6-Chloro-5-methylnicotinic acid ≥97%
6-Chloro-5-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.581 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17214192
CAS Number: 66909-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR