5-Chloro-2-fluoronicotinaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
5-Chloro-2-fluoro-3-formylpyridine
14133-25G
14133-5G
76925-090EA
616.39
USD
76925-090
76925-092
5-Chloro-2-fluoronicotinaldehyde ≥97%
5-Chloro-2-fluoronicotinaldehyde
Formula:
C₆H₃ClFNO MW: 159.55 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04972395 CAS Number: 882679-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...