4-Formyl-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

14116-1G 14116-25G 14116-500MG 14116-5G
76925-076EA 125.19 USD
76925-076 76925-078 76925-080 76925-082
4-Formyl-3-methylbenzoic acid ≥97%
4-Formyl-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₃
Storage Temperature: Refrigerator
MDL Number: MFCD20660431
CAS Number: 24078-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR