5-Ethoxypyridine-3-boronic acid ≥95%

Supplier: AOBChem USA

Warning

14102-1G 14102-25G 14102-500MG 14102-5G
76925-046EA 180.36 USD
76925-046 76925-048 76925-050 76925-052
5-Ethoxypyridine-3-boronic acid ≥95%
5-Ethoxypyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₃
MW: 166.97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375005
CAS Number: 1224436-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR