3-(2-Chlorophenylcarbamoyl)-4-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-3-(2-chloro-phenylcarbamoyl)phenylboronic acid

14087-1G 14087-25G 14087-5G
76925-032EA 64.72 USD
76925-032 76925-034 76925-036
3-(2-Chlorophenylcarbamoyl)-4-fluorophenylboronic acid ≥97%
3-(2-Chlorophenylcarbamoyl)-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀BClFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11856003
CAS Number: 1451393-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR