5-Bromo-2-chloroisopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

14063-1G 14063-25G 14063-500MG 14063-5G
76925-020EA 233.4 USD
76925-020 76925-022 76925-024 76925-026
5-Bromo-2-chloroisopropylbenzene ≥97%
5-Bromo-2-chloroisopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrCl
Storage Temperature: Ambient
MDL Number: MFCD11845957
CAS Number: 90350-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR