(4-Chloro-3-ethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-3-ethylphenylboronic acid

14042-1G 14042-25G 14042-500MG 14042-5G
76924-996EA 141.1 USD
76924-996 76924-998 76925-000 76925-002
(4-Chloro-3-ethylphenyl)boronic acid ≥97%
(4-Chloro-3-ethylphenyl)boronic acid
Formula: C₈H₁₀BClO₂
MW: 184.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566599
CAS Number: 918810-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR