4-Fluoro-3-(4-morpholinylcarbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-3-(morpholine-4-carbonyl)phenylboronic acid

14017-1G 14017-25G 14017-500MG 14017-5G
76924-960EA 138.98 USD
76924-960 76924-962 76924-964 76924-966
4-Fluoro-3-(4-morpholinylcarbonyl)phenylboronic acid ≥97% research grade
4-Fluoro-3-(4-morpholinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₃BFNO₄
Storage Temperature: Ambient
MDL Number: MFCD08235048
CAS Number: 874219-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR