4-Fluoro-3-(1-pyrrolidinylcarbonyl)phenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-3-(1-pyrrolidinylcarbonyl)benzeneboronic acid, 4-Fluoro-3-(pyrrolidine-1-carbonyl)phenylboronic acid

14015-1G 14015-25G 14015-5G
76924-946EA 96.55 USD
76924-946 76924-948 76924-950
4-Fluoro-3-(1-pyrrolidinylcarbonyl)phenylboronic acid ≥95% research grade
4-Fluoro-3-(1-pyrrolidinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₃BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08235050
CAS Number: 874219-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR