4-Fluoro-3-(methylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Fluoro-3-(methylcarbamoyl)phenyl)boronic acid, 4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid

14013-25G 14013-5G
76924-934EA 630.18 USD
76924-934 76924-936
4-Fluoro-3-(methylcarbamoyl)phenylboronic acid ≥97%
4-Fluoro-3-(methylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD08235038
CAS Number: 874219-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR