5-Chloro-3-fluoropyridin-2ylhydrazine ≥97%

Supplier: AOBChem USA

Warning

13789-1G 13789-25G 13789-5G
76924-896EA 79.57 USD
76924-896 76924-898 76924-900
5-Chloro-3-fluoropyridin-2ylhydrazine ≥97%
5-Chloro-3-fluoropyridin-2ylhydrazine

Precaución: for Research Use Only

Formula: C₅H₅ClFN₃
Storage Temperature: Refrigerator
MDL Number: MFCD09972183
CAS Number: 248255-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR