4-Formyl-3-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Formyl-3-(trifluoromethyl)benzeneboronic acid

13438-1G 13438-25G 13438-500MG 13438-5G
76924-854EA 143.23 USD
76924-854 76924-856 76924-858 76924-860
4-Formyl-3-(trifluoromethyl)phenylboronic acid ≥97%
4-Formyl-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₆BF₃O₃
MW: 217.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037498
CAS Number: 1072944-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR