5-Carboxy-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Borono-4-methoxybenzoic acid

13338-25G 13338-5G
76924-842EA 495.45 USD
76924-842 76924-844
5-Carboxy-2-methoxyphenylboronic acid ≥97%
5-Carboxy-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₅
MW: 195.9700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203261
CAS Number: 730971-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR