5-Acetamido-2-nitrophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Acetamido-2-nitro)benzeneboronicacid

13229-1G 13229-25G 13229-500MG 13229-5G
76924-828EA 124.13 USD
76924-828 76924-830 76924-832 76924-834
5-Acetamido-2-nitrophenylboronic acid ≥97%
5-Acetamido-2-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD06659829
CAS Number: 78887-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR