5-Bromo-2-(pyrrolidin-1-ylcarbonyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

12762-1G 12762-25G 12762-5G
76924-796EA 66.84 USD
76924-796 76924-798 76924-800
5-Bromo-2-(pyrrolidin-1-ylcarbonyl)pyridine ≥97%
5-Bromo-2-(pyrrolidin-1-ylcarbonyl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09801066
CAS Number: 742085-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR