5-Bromo-N-methylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

12757-25G 12757-5G
76924-788EA 289.63 USD
76924-788 76924-790
5-Bromo-N-methylnicotinamide ≥97%
5-Bromo-N-methylnicotinamide
Formula: C₇H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD01788369
CAS Number: 153435-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR