5-tert-Butyl-2-methoxybenzeneboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (5-(tert-Butyl)-2-methoxyphenyl)boronic acid, 5-(tert-Butyl)-2-methoxyphenylboronic acid

12704-1G 12704-25G 12704-5G
76924-778EA 102.91 USD
76924-778 76924-780 76924-782
5-tert-Butyl-2-methoxybenzeneboronic acid ≥97% research grade
5-tert-Butyl-2-methoxybenzeneboronic acid
Formula: C₁₁H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD06201034
CAS Number: 128733-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR