4-(Methanesulfonamido)phenylboronic acid pinacol ester ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-[4-(Methanesulphonylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-(Methylsulphonyl)-4-(4,4,5,5-tetraMethyl-1,3,2-dioxaborolan-2-yl)benzenamine, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanesulphonamide, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-phenyl)methanesulphonamide, 4-(Methanesulphonylamino)phenylboronic acid Pinacol ester, 4-Methanesulphonylaminophenylboronic acid, pinacol ester, N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulphonamide, 4-(Methylsulphonamido)phenylboronic acid pinacol ester

12491-25G 12491-5G
76924-766EA 418 USD
76924-766 76924-768
4-(Methanesulfonamido)phenylboronic acid pinacol ester ≥95% research grade
4-(Methanesulfonamido)phenylboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₄S
Storage Temperature: Ambient
MDL Number: MFCD05663876
CAS Number: 616880-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR