4-Hydroxy-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

12380-25G 12380-5G
76924-756EA 410.57 USD
76924-756 76924-758
4-Hydroxy-2-methylphenylboronic acid ≥97%
4-Hydroxy-2-methylphenylboronic acid
Formula: C₇H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD03788424
CAS Number: 493035-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR