4-Phenylcarbamoylbenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Phenylaminocarbonyl)benzeneboronic acid, 4-Phenylaminocarbonylphenylboronic acid, (4-(Phenylcarbamoyl)phenyl)boronic acid

12282-25G 12282-5G
76924-750EA 310.85 USD
76924-750 76924-752
4-Phenylcarbamoylbenzeneboronic acid ≥97%
4-Phenylcarbamoylbenzeneboronic acid
Formula: C₁₃H₁₂BNO₃
MW: 241.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115683
CAS Number: 330793-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR