5-Chloro-3,4'-bipyridine ≥97%

Supplier: AOBChem USA

Warning

122111-1G 122111-500MG 122111-5G 122111-25G
76924-736EA 1012.1 USD
76924-736 76924-738 76924-740 77635-422
5-Chloro-3,4'-bipyridine ≥97%
5-Chloro-3,4'-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂
MW: 190.629 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15475280
CAS Number: 1214337-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR