4-Methyl-2-thiopheneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Methylthiophen-2-yl)boronic acid, 4-Methylthiophene-2-boronic acid

12159-25G 12159-5G
76924-700EA 233.4 USD
76924-700 76924-702
4-Methyl-2-thiopheneboronic acid ≥97%
4-Methyl-2-thiopheneboronic acid

Precaución: for Research Use Only

Formula: C₅H₇BO₂S
MW: 141.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01319040
CAS Number: 162607-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR