4-Isopropoxy-2-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Isopropoxy-2-trifluoromethylphenylboronic acid

12131-1G 12131-25G 12131-5G
76924-686EA 97.61 USD
76924-686 76924-688 76924-690
4-Isopropoxy-2-(trifluoromethyl)phenylboronic acid ≥97%
4-Isopropoxy-2-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD04972373
CAS Number: 313545-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR