4-Bromo-2-fluoro-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

120953-1G 120953-25G 120953-500MG 120953-5G
76924-668EA 360.71 USD
76924-668 76924-670 76924-672 76924-674
4-Bromo-2-fluoro-5-methoxybenzoic acid ≥97%
4-Bromo-2-fluoro-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD29931556
CAS Number: 1620677-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR