(5-Fluoro-2-propoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

12006-25G 12006-5G
76924-644EA 418 USD
76924-644 76924-646
(5-Fluoro-2-propoxyphenyl)boronic acid ≥97%
(5-Fluoro-2-propoxyphenyl)boronic acid
Formula: C₉H₁₂BFO₃
MW: 198.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664233
CAS Number: 480438-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR