4-Bromo-5-nitroisoquinoline ≥97% research grade

Supplier: AOBChem USA

Warning

119257-1G 119257-25G 119257-5G
76924-628EA 62.6 USD
76924-628 76924-630 76924-632
4-Bromo-5-nitroisoquinoline ≥97% research grade
4-Bromo-5-nitroisoquinoline
Formula: C₉H₅BrN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00234471
CAS Number: 58142-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR