5-Formylpyridine-3-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinaldehyde

11595-1G 11595-25G 11595-5G
76924-618EA 62.6 USD
76924-618 76924-620 76924-622
5-Formylpyridine-3-boronic acid pinacol ester ≥97% research grade
5-Formylpyridine-3-boronic acid pinacol ester
Formula: C₁₂H₁₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD04038759
CAS Number: 848093-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR